Learn More
Bio X Cell IgG1 Fc, 34 kDa, 5 MG
Supplier: Bio X Cell BE0096R005MG
This recombinant human IgG1 Fc is the Fc fragment of human IgG1, commonly used as an isotype control for human IgG1 antibodies and fusion proteins. It has a molecular mass of approximately 34 kDa and a purity of ≥95%.
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives
- Endotoxin levels are ≤1EU/mg (≤0.001EU/μg)
- Sterility ensured by 0.2 μm filtration
- Produced in an animal-free facility and purified via Protein A
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.