Learn More
Proteintech Group Inc Recomb. Human IFN 2A 10ug
SDP
Supplier:  Proteintech Group Inc HZ106610UG
Encompass_Preferred
Animal-free Recombinant Human IFN alpha 2A is expressed in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 16 kDa. It is a type 1 interferon and is also known as leukocyte interferon. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation which contributes to stability in cell growth media and other applications. The purity is greater than 95%.
Catalog No. 50-197-3865