Learn More
Proteintech Group Inc Recomb. Human IFN 2B 1000ug
SDP
Supplier:  Proteintech Group Inc HZ10721000UG
Encompass_Preferred
Animal-free Recombinant Human IFN alpha 2B is expressed in human 293 cells as a monomeric glycoprotein with an apparent molecular mass of 16 kDa. This cytokine is produced in a serum-free, chemically defined media. Production in human 293 cells offers authentic glycosylation which contributes to stability in cell growth media and other applications. The purity is greater than 95%.
Catalog No. 50-197-3868