Learn More
ILEX LIFE SCIENCES LLC Recombinant Human Insulin Protein (E. coli), 250 mg
SDP
Supplier:  ILEX LIFE SCIENCES LLC A32700250
Encompass_Preferred
Recombinant Human Insulin Protein, produced in E. coli, is a two chain, non-glycosylated polypeptide hormone containing 51 amino acids and has a 5807 Dalton molecular mass. CAS REGISTRY NO.: 11061-68-0. PURITY: Purity greater than 98.0% as determined by RP-HPLC analysis. BIOLOGICAL ACTIVITY: The biological activity was determined to be 28 units/mg. FORMULATION: Sterile-filtered white lyophilized (freeze-dried) powder. The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. SUMMARY: Insulin is a peptide hormone produced by beta cells of the pancreatic islets encoded in humans by the INS gene. Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver (UniProtKb_P01308). Recombinant Human Insulin is a commonly used media supplement in mammalian cell culture.
Catalog No. 50-233-9503