Learn More
Abcam Recombinant Human IRF5 protein, 10UG
Supplier:  Abcam AB17302410UG
Encompass_Preferred
Recombinant Human IRF5 protein is a Human Full Length protein, in the 1 to 498 aa range, expressed in Escherichia coli, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Interferon regulatory factor 5, IRF-5, IRF5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1970165