Learn More
Abcam Recombinant human PKR protein, 5UG
Supplier:  Abcam AB858385UG
Encompass_Preferred
Recombinant human PKR protein is a Human Fragment protein, in the 252 to 551 aa range, expressed in Baculovirus infected Sf9, with >80% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: PKR, PRKR, EIF2AK2, Eukaryotic translation initiation factor 2-alpha kinase 2, Interferon-inducible RNA-dependent protein kinase, P1/eIF-2A protein kinase, Protein kinase RNA-activated, Tyrosine-protein kinase EIF2AK2, p68 kinase, eIF-2A protein kinase 2, Protein kinase R
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2306629