Learn More
Abcam Recombinant Human PPP2R2A protein, 10UG
Supplier:  Abcam AB15917210UG
Encompass_Preferred
Recombinant Human PPP2R2A protein is a Human Full Length protein, in the 1 to 447 aa range, expressed in Wheat germ and suitable for ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform, PP2A subunit B isoform B55-alpha, PP2A subunit B isoform PR55-alpha, PP2A subunit B isoform R2-alpha, PP2A subunit B isoform alpha, PPP2R2A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2134843