Learn More
Proteintech Group Inc Recomb. Human TGF beta 2 5ug
SDP
Supplier:  Proteintech Group Inc HZ10925UG
Encompass_Preferred
Animal-free Recombinant Human TGF beta 2HuXp is expressed from human 293 cells as a mature disulfide-linked homodimer with apparent molecular mass of 25 kDa. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. The cytokine is greater than 95% pure.
Catalog No. 50-197-3880