Learn More
Abcam Recombinant human TGF beta 1 protein (Active), 20UG
Supplier:  Abcam AB11615020UG
Encompass_Preferred
Recombinant human TGF beta 1 protein (Active) is a Human Full Length protein, in the 279 to 390 aa range, expressed in CHO, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, FuncS.
Protein/ Peptide/ Enzyme Synonyms: TGFB, TGFB1, Transforming growth factor beta-1 proprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2174818