Learn More
Abcam Recombinant human TYK2 protein, 10UG
Supplier:  Abcam AB12553910UG
Encompass_Preferred
Recombinant human TYK2 protein is a Human Fragment protein, in the 442 to 1187 aa range, expressed in Baculovirus infected Sf9, with >70% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Non-receptor tyrosine-protein kinase TYK2, TYK2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2066044