Learn More
Cosmo Bio Usa Inc RECOMB SALMONELLA TYPHIM 20UG
SDP
Catalog No. NC3358245
Encompass
Recombinant Salmonella typhimurium Phase 2 flagellin /fljB/ partial Biotinylated 20ug.Purity greater than 85 percent as determined by SDS-PAGE.Target Names fijBSpecies Salmonella typhimurium strain LT2/SGSC1412/ATCC 700720Source Ecoli
Catalog No. NC3358245 Supplier Cosmo Bio Usa Inc Supplier No. CSBEP345218SXBB20
Recombinant Salmonella typhimurium Phase 2 flagellin /fljB/ partial Biotinylated 20ug.Purity greater than 85 percent as determined by SDS-PAGE.Target Names fijBSpecies Salmonella typhimurium strain LT2/SGSC1412/ATCC 700720Source Ecoli