Learn More
Bio X Cell IgG1 LALA-PG, Anti-Hen Egg Lysozyme, 1 MG
Supplier: Bio X Cell CP149R001MG
This human IgG1 isotype control antibody reacts with hen egg lysozyme and has low or no specific binding to any human sample. The L234A, L235A, and P329G (LALA-PG) mutations decrease effector functions by disrupting the Fc/Fcγ receptor interface.
- Isotype: human IgG1
- Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
- Conjugation: unconjugated
- Immunogen: hen egg lysozyme (HEL)
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg), determined by LAL assay
- Aggregation: <5%, determined by SEC
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from CHO cell supernatant, animal-free facility
- Purification: protein A
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.