Learn More
Bio X Cell RecombiMAb, Mouse Ly6G, 1A8, IgG2a, 100 MG
Supplier: Bio X Cell CP129R100MG
The 1A8-CP129 monoclonal antibody is a recombinant Fc-engineered chimeric version of the original 1A8 antibody, designed for efficient in vivo Ly6G targeted cell depletion. Its mouse IgG2a isotype reduces immunogenicity in mouse models compared to xenogenic antibodies. This antibody specifically reacts with mouse Ly6G, a key cell surface marker for myeloid lineage cells.
- Reacts specifically with mouse Ly6G
- Fc-engineered chimeric antibody
- Mouse IgG2a isotype, kappa light chain
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤0.5 EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Applications include in vivo neutrophil and MDSC depletion, Western Blot, Immunofluorescence, Immunohistochemistry, and Flow cytometry
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.