Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell RecombiMAb Anti-Mouse PD-1, Clone 29F.1A12, 1 MG

Catalog No. NC3252800
Encompass_Preferred

The 29F.1A12™-CP178 monoclonal antibody is a recombinant chimeric antibody that reacts with mouse PD-1 (CD279). Its constant region has been switched from rat IgG2a to mouse IgG1, while retaining identical variable domain sequences. This antibody is designed to target PD-1, a cell surface receptor involved in inhibiting T-cell activation and playing key roles in peripheral tolerance.

  • Purified from CHO cell supernatant in an animal-free facility
  • Purification by protein G
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Isotype: mouse IgG1, κ
  • Molecular weight: 150 kDa
  • Unconjugated
  • Endotoxin ≤0.5 EU/mg (≤0.0005 EU/µg)
  • Purity ≥95% by SDS-PAGE
  • Sterility by 0.2 µm filtration
  • Reported applications include in vivo blocking of PD-1/PD-L signaling and in vitro PD-1 neutralization

Catalog No. NC3252800 Supplier Bio X Cell Supplier No. CP178R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 29F.1A12™-CP178 monoclonal antibody is a recombinant chimeric antibody that reacts with mouse PD-1 (CD279). Its constant region has been switched from rat IgG2a to mouse IgG1, while retaining identical variable domain sequences. This antibody is designed to target PD-1, a cell surface receptor involved in inhibiting T-cell activation and playing key roles in peripheral tolerance.

  • Purified from CHO cell supernatant in an animal-free facility
  • Purification by protein G
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Isotype: mouse IgG1, κ
  • Molecular weight: 150 kDa
  • Unconjugated
  • Endotoxin ≤0.5 EU/mg (≤0.0005 EU/µg)
  • Purity ≥95% by SDS-PAGE
  • Sterility by 0.2 µm filtration
  • Reported applications include in vivo blocking of PD-1/PD-L signaling and in vitro PD-1 neutralization

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.