Learn More
Bio X Cell Human IgG1 (N297A) Isotype, Anti-HEL, 1 MG
Supplier: Bio X Cell CP171R001MG
This recombinant human IgG1 (N297A) isotype control antibody, produced in CHO cells, reacts with hen egg lysozyme and exhibits low or no specific binding to human samples. The N297A mutation prevents glycosylation, markedly decreasing Fc receptor binding.
- Isotype: Human IgG1, κ
- Immunogen: Hen egg lysozyme (HEL)
- Mutations: N297A
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤0.5 EU/mg (≤0.0005 EU/μg) determined by LAL assay
- Aggregation: <5% determined by SEC
- Purity: ≥95% determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Purified from CHO cell supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Conjugation: Unconjugated
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.