Learn More
Bio X Cell RecombiMAb mouse IgG2a (D265A) isotype control, anti-hen egg lysozyme - 5 MG (Catalog: CP150-R005MG), 5 MG
Supplier: Bio X Cell CP150R005MG
Bio X Cell RecombiMAb mouse IgG2a (D265A) isotype control (anti-hen egg lysozyme) is a recombinant monoclonal antibody supplied as a purified solution for use as an isotype-matched control in in vivo and in vitro experiments. The antibody carries an Fc D265A mutation and is supplied at 5 mg with >95% purity; store at 4C and do not freeze.
- Recombinant monoclonal isotype control antibody.
- Isotype: mouse IgG2a with D265A Fc mutation.
- Target specificity: anti-hen egg lysozyme; low or no specific binding to mouse samples.
- Purity greater than 95% as determined by SDS-PAGE.
- Approximate molecular weight 150 kDa.
- Supplied as a purified antibody solution, 5 mg.
- Storage: store at 4C; do not freeze.
- Shipped cold to maintain 4C during transport.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.