Learn More
Takara Bio Recombinant DNase I (RNase-free), 1,000 Units
SDP
Catalog No. 50443716
Encompass
Recombinant DNase I (RNase-free) is an endonuclease that catalyzes, to the same degree, the random degradation of both single- and double-stranded DNA and produces 5'-P terminal oligonucleotides. This enzyme does not exhibit protease activity. This enzyme is stable around its optimum neutral pH range and is suitable for RNA preparation at neutral pH. 1000 Units
Catalog No. 50-443-716 Supplier Takara Bio Supplier No. 2270A
Recombinant DNase I (RNase-free) is an endonuclease that catalyzes, to the same degree, the random degradation of both single- and double-stranded DNA and produces 5'-P terminal oligonucleotides. This enzyme does not exhibit protease activity. This enzyme is stable around its optimum neutral pH range and is suitable for RNA preparation at neutral pH. 1000 Units