Learn More
Takara Bio RECOMBINA DNASE I (RNASE-FREE)
SDP
Catalog No. 50443717
Encompass
Enzyme, Modifying; Recombinant DNase I (RNase-free); Takara; 5000U; 5U/uL; Source: Expressed in non-animal host carrying plasmid encodes the gene of bovine pancreas DNase I; Storage: -20 deg.C; Buffers included
Catalog No. 50-443-717 Supplier Takara Bio Supplier No. 2270B
Enzyme, Modifying; Recombinant DNase I (RNase-free); Takara; 5000U; 5U/uL; Source: Expressed in non-animal host carrying plasmid encodes the gene of bovine pancreas DNase I; Storage: -20 deg.C; Buffers included