Learn More
R&D Systems™ Recombinant A. Muciniphila M60 domain-containing peptidase
Description
Specifications
Specifications
| Accession Number | WP_012419679.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Molecular Weight (g/mol) | MolecularWeight-observed: 52-57 kDa, under reducing conditions, MolecularWeight-theroretical: 56 kDa |
| Quantity | 20 μg |
| Source | E. coli-derived akkermansia muciniphila M60 domain-containing protein Ala21-Glu506 with an N-terminal Met and C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Akkermansia muciniphila ATCC BAA-835 Peptidase M60, Akkermansia muciniphila ATCC BAA-835 Peptidase M60 domain-containing protein, Amuc_0627, conserved hypothetical protein [Akkermansia mucini, conserved hypothetical protein [Akkermansia muciniphila ATCC BAA-835], M60 family metallopeptidase [Akkermansia muciniphi, M60 family metallopeptidase [Akkermansia muciniphila] |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.