Learn More
New England Biolabs, Inc. Recombinant Albumin, Molecular Biology Grade - 250 mg
SDP
Catalog No. 5000460443
Encompass
Need to avoid BSA or albumin from an animal source? Recombinant Albumin, Molecular Biology Grade, is a non-bovine derived albumin that can serve as an alternative to Bovine Serum Albumin (BSA). Like BSA, it has been shown to prevent adhesion of enzymes to reaction tubes and pipette surfaces. It also stabilizes some proteins during incubation. Choose Recombinant Albumin, when there is a need to avoid BSA. Please inquire about custom formulations with Recombinant Albumin.
  • Recombinant non-bovine derived albumin
  • Equivalent activity to Bovine Serum Albumin (BSA)
  • Available in convenient large size
  • Availabe in a convenient 250mg format for added convenience
Catalog No. 50-004-60443 Supplier New England Biolabs, Inc. Supplier No. B9200L
Need to avoid BSA or albumin from an animal source? Recombinant Albumin, Molecular Biology Grade, is a non-bovine derived albumin that can serve as an alternative to Bovine Serum Albumin (BSA). Like BSA, it has been shown to prevent adhesion of enzymes to reaction tubes and pipette surfaces. It also stabilizes some proteins during incubation. Choose Recombinant Albumin, when there is a need to avoid BSA. Please inquire about custom formulations with Recombinant Albumin.
  • Recombinant non-bovine derived albumin
  • Equivalent activity to Bovine Serum Albumin (BSA)
  • Available in convenient large size
  • Availabe in a convenient 250mg format for added convenience