Learn More
Creative Biomart Alfimeprase, Recombinant fibrolase
Supplier:  Creative Biomart THP0203
Encompass_Preferred
The product is a recombinant analog of fibrolase. Fibrolase is a zinc-containing metalloproteinase isolated from the venom of the southern copperhead snake (Agkistrodon contortrix contortrix). It is a small protein that contains 203 residues (Randolph et al. 1992).
Catalog No. 50-208-4254