Learn More
Sino Biological Recombinant Anti-ACE2 Antibody, Rabbit Monoclonal 20 µL
Catalog No. 5000786014
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ACE2 (rR ACE2; Catalog#80031-R08H; Q5EGZ1; Met 1-Thr 740).
Catalog No. 50-007-86014 Supplier Sino Biological Supplier No. 80031R00120
This antibody was obtained from a rabbit immunized with purified, recombinant Rat ACE2 (rR ACE2; Catalog#80031-R08H; Q5EGZ1; Met 1-Thr 740).