Learn More
Sino Biological Recombinant Anti-B7-2/CD86 Antibody, Rabbit Monoclonal 100 µL
Catalog No. 5000786138
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat B7-2/CD86 (rR B7-2/CD86; Catalog#80025-R08H; NP_064466.1; Met1-Ile250).
Catalog No. 50-007-86138 Supplier Sino Biological Supplier No. 80025R053100
This antibody was obtained from a rabbit immunized with purified, recombinant Rat B7-2/CD86 (rR B7-2/CD86; Catalog#80025-R08H; NP_064466.1; Met1-Ile250).