Learn More
Sino Biological Recombinant Anti-BCMA Antibody, Rabbit Monoclonal 100 µL
Catalog No. 5000751308
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human BCMA (rh BCMA; Catalog#10620-H08H; NP_001183.2; Met1-Ala54).
Catalog No. 50-007-51308 Supplier Sino Biological Supplier No. 10620R054100
This antibody was obtained from a rabbit immunized with purified, recombinant Human BCMA (rh BCMA; Catalog#10620-H08H; NP_001183.2; Met1-Ala54).