Learn More
Abcam Anti-CD31 antibody [RM1006] - BSA and Azide free
Supplier:  Abcam AB282746100UG
Encompass_Preferred
Rabbit recombinant multiclonal [RM1006] to CD31 - BSA and Azide free
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2879904