Learn More
Abcam Anti-EHMT2/G9A antibody [EPR18894], ChIP Grade, 100 μL
Supplier: Abcam AB185050100UL
This product is a recombinant anti-EHMT2/G9A antibody (clone EPR18894), designed for reliable detection of the EHMT2/G9A protein in various applications. The antibody is knockout validated and offers high specificity, making it ideal for epigenetic research. Its rabbit monoclonal format ensures excellent consistency across different batches and minimized background interference.
- ChIP-grade antibody suitable for multiple applications
- Recombinant monoclonal format providing batch-to-batch consistency
- Validated for Western Blot, Flow Cytometry, and Immunoprecipitation
- Reacts with human, mouse, and rat proteins
- Knockout validated for high specificity in EHMT2 analysis
- High performance in immunocytochemistry and immunohistochemistry
- Concentration of 0.865 mg/ml
- Available in 100 μL or 1 mL quantities
- Cited in over 30 peer-reviewed publications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.