Learn More
Sino Biological Recombinant Anti-EPCR Antibody (FITC), Rabbit Monoclonal 100 Tests
Catalog No. 5000785910
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Epcr/PROCR (rM Epcr/PROCR; Catalog#50874-M08H; Q64695; Met1-Ser214) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-007-85910 Supplier Sino Biological Supplier No. 50874R011F100
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Epcr/PROCR (rM Epcr/PROCR; Catalog#50874-M08H; Q64695; Met1-Ser214) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.