Learn More
Sino Biological Recombinant Anti-Kanamycin Antibody, Rabbit Monoclonal 20 µg
Catalog No. 5000785536
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, Kanamycin conjugated to BSA.
Catalog No. 50-007-85536 Supplier Sino Biological Supplier No. 68257R001720
This antibody was obtained from a rabbit immunized with purified, Kanamycin conjugated to BSA.