Learn More
Sino Biological Recombinant Anti-Nectin-1/PVRL1 Antibody (Biotin), Rabbit Monoclonal 20 µg
Catalog No. 5000786591
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD111 / Nectin-1 / PVRL1 (Catalog#80244-R08H; XP_236210.3; Met1-Ala354), and then biotinylated.
Catalog No. 50-007-86591 Supplier Sino Biological Supplier No. 80244R004B20
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD111 / Nectin-1 / PVRL1 (Catalog#80244-R08H; XP_236210.3; Met1-Ala354), and then biotinylated.