Learn More
Sino Biological Recombinant Anti-Serpin A1/Alpha-1-antitrypsin Antibody (Biotin), Rabbit Monoclonal 100 µg
Catalog No. 5000750506
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinA1 (rh SerpinA1; Catalog#10306-H08H; NP_000286.3; Met1-Lys418), and then biotinylated.
Catalog No. 50-007-50506 Supplier Sino Biological Supplier No. 10306R005B100
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinA1 (rh SerpinA1; Catalog#10306-H08H; NP_000286.3; Met1-Lys418), and then biotinylated.