Learn More
Sino Biological Recombinant Anti-Whitlow 218 linker Antibody (APC), Rabbit Monoclonal 100 Tests
Catalog No. 5000749807
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the Whitlow 218 linker, and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-007-49807 Supplier Sino Biological Supplier No. 111774R0013A100
This antibody was obtained from a rabbit immunized with purified, synthetic peptide corresponding to the Whitlow 218 linker, and conjugated with APC under optimum conditions, the unreacted APC was removed.