Learn More
Active Motif Recombinant BRD4 (333-460) protein
SDP
Supplier:  Active Motif 31446
Encompass
The peptide corresponding to amino acids 333-460 that contains the bromodomain sequences of BRD4 (accession number NP_490597.1) was expressed in E. coli and contains an N-terminal His tag and C-terminal FLAG tag with an observed molecular weight of 20.9 kDa. Bromodomain-containing protein 4 (BRD4) belongs to the BET subclass of proteins, which are characterized by two N-terminal bromodomains and one ET (Extra Terminal) domain. BRDs associate with chromatin through their bromodomains that recognize acetylated histone lysine residues. Bromodomains function as ïreaders' of these epigenetic histone marks and regulate chromatin structure and gene expression by linking associated proteins to the acetylated nucleosomal targets. The ET domain functions as a protein binding motif and exerts atypical serine-kinase […] For more information, please visit: https://www.activemotif.com/catalog/details/31446.
Catalog No. NC1845631
Edge