Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant canine interleukin-21 | >95.0% | 50 UG
SDP

Catalog No. 5000293722
Encompass_Preferred

Recombinant canine interleukin-21 (IL-21) is a tag-free, lyophilized cytokine produced in E. coli representing residues H18-S146 (Accession Q6L7I9). Supplied as a 50 μg vial with >95% purity by reducing SDS-PAGE, it is intended for in vitro research and bioactivity assays. Predicted molecular mass is ~15 kDa and bioactivity is reported with an ED50 of 7.767 ng/mL in an NK-92 IFN-γ induction assay. Store lyophilized at -20°C and aliquot for long-term storage.

  • Tag-free, recombinant canine IL-21 for species-relevant studies.
  • Expressed in E. coli for efficient production.
  • Represents residues H18-S146 of the native sequence.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 50 μg lyophilized aliquot for convenience.
  • Predicted molecular mass of ~15 kDa for gel identification.
  • Bioactivity verified with ED50 7.767 ng/mL in NK-92 IFN-γ assay.
  • Store at -20°C; aliquot and freeze at -80°C for extended stability.

Catalog No. 50-002-93722 Supplier Medchemexpress LLC Supplier No. HYP7481950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine interleukin-21 (IL-21) is a tag-free, lyophilized cytokine produced in E. coli representing residues H18-S146 (Accession Q6L7I9). Supplied as a 50 μg vial with >95% purity by reducing SDS-PAGE, it is intended for in vitro research and bioactivity assays. Predicted molecular mass is ~15 kDa and bioactivity is reported with an ED50 of 7.767 ng/mL in an NK-92 IFN-γ induction assay. Store lyophilized at -20°C and aliquot for long-term storage.

  • Tag-free, recombinant canine IL-21 for species-relevant studies.
  • Expressed in E. coli for efficient production.
  • Represents residues H18-S146 of the native sequence.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 50 μg lyophilized aliquot for convenience.
  • Predicted molecular mass of ~15 kDa for gel identification.
  • Bioactivity verified with ED50 7.767 ng/mL in NK-92 IFN-γ assay.
  • Store at -20°C; aliquot and freeze at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.