Learn More
Kactus Recombinant Canine LRRC15/LIB Protein is expressed from HEK293 with His tag at the C-Terminus.It contains Tyr47-Gly565.
Supplier:  Kactus LRRDM415100UG
Encompass_Preferred
Canine LRRC15/LIB Protein (LRR-DM415). Endotoxin is less than 1 EU per μg by the LAL method. The purity of this protein is determined to be > 95% by Bis-Tris PAGE and > 95% by HPLC.
Catalog No. 50-004-63884