Learn More
Creative Biomart Alvircept sudotox, Recombinant CD4 Protein, sCD4-PE40
Supplier:  Creative Biomart THP0289
Encompass_Preferred
Alvircept sudotox is a form of recombinant CD4 derived from Pneumonas aeruginosa exotoxin A, or 'PE40, which has a size of 59,187 daltons and is an anti-viral agent.
Catalog No. 50-208-4339