Learn More
Creative Biomart Nonacog beta pegol, Recombinant coagulation factor IX
Supplier:  Creative Biomart THP0214
Encompass_Preferred
The product is a recombinant coagulation factor IX derivative. It is produced without animal-derived materials and with an attached 40kDa polyethylene glycol (PEG) molecule for peptide activation by a site-directed glycoPEGylation. Once activated, the activation molecule with PEG are cleaved to leave the activated factor IX (Factor IXa).
Catalog No. 50-208-4265