Learn More
Abcam Recombinant Cow Mannan Binding Lectin/MBL protein (Tagged), 10UG
Catalog No. 502704135
Encompass_Preferred
Recombinant Cow Mannan Binding Lectin/MBL protein (Tagged) is a Cow Full Length protein, in the 21 to 249 aa range, expressed in Mammalian, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Mannose-binding protein C, MBP-C, Mannan-binding protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4135 Supplier Abcam Supplier No. AB26789510UG
Recombinant Cow Mannan Binding Lectin/MBL protein (Tagged) is a Cow Full Length protein, in the 21 to 249 aa range, expressed in Mammalian, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Mannose-binding protein C, MBP-C, Mannan-binding protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement