Learn More
Biomatik Corporation Recombinant Cricetulus griseus Peroxiredoxin-1 (PRDX1)
Supplier:  Biomatik Corporation RPC23613
Encompass_Preferred
Recombinant Cricetulus griseus Peroxiredoxin-1 (PRDX1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus). Target Name: PRDX1. Target Synonyms: Thioredoxin peroxidase 2TPX-2. Accession Number: Q9JKY1. Expression Region: 2~199aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5748