Learn More
R&D Systems™ Recombinant Cyno Lymphotoxin beta R/TNFRSF3 His Protein
Description
Specifications
Specifications
| Accession Number | XP_005569951.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4055 |
| Molecular Weight (g/mol) | M.W.-observed: 29-41 kDa, under reducing conditions., M.W.-theroretical: 26 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Lymphotoxin beta R/TNFRSF3 protein Ser28-Met227, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.