Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus 4-1BB/TNFRSF9 Fc Chimera Protein

Catalog No. 93244B100
Change view
Click to view available options
Quantity: 100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
93-244-B100 Quantity: 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-244-B100 Supplier R&D Systems™ Supplier No. 93244B100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey 4-1BB/TNFRSF9/CD137 Fc Chimera is immobilized at 0.05 μg/mL (100 μL/well), Recombinant Human 4-1BB Ligand/TNFSF9 (Catalog # 2295-4L ) binds with an ED50 of 0.25-1.25 ng/mL.

Order Info

Ships at ambient temperature

Specifications

Accession Number XP_005544945
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3604
Molecular Weight (g/mol) M.W. (Observed): 56-66 kDa, reducing conditions; M.W. (theoretical): 44 kDa
Name 4-1BB/TNFRSF9 Fc Chimera Recombinant Protein
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey 4-1 BB/TNFRSF9/CD137 protein Cynomolgus Monkey 4-1 BB/TNFRSF9 (Leu24-Gln186) (N-terminus) Accession # XP_005544945 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Common Name 4-1BB ligand receptor; 41BB; 4-1BB; CD137 antigen; CD137; CD137MGC2172; CDw137; FLJ43501; ILA; ILAhomolog of mouse 4-1BB; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; receptor protein 4-1BB; T cell antigen
Biological Activity 0.25 to 1.25ng/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein 4-1 BB/TNFRSF9/CD137
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.