Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus CD200R1 His-tag Protein

Catalog No. 10055CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-055-CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-055-CD050 Supplier R&D Systems™ Supplier No. 10055CD050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Cynomolgus CD200 R1 His-tag Protein is derived from HEK293. The Recombinant Cynomolgus CD200 R1 His-tag Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number XP_005548208
For Use With (Application) Bioactivity
Gene ID (Entrez) 131450
Molecular Weight (g/mol) 28 kDa
Name CD200R1
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived cynomolgus monkey CD200 R1 protein Ala27-Leu267,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD200 R1, CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, MOX2R, OX2RCD200 cell surface glycoprotein receptor
Specific Reactivity CD200R1
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 1 mg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.