Learn More
R&D Systems™ Recombinant Cynomolgus Fc gamma RIII/CD16 Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | NP_001270121 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl |
| Gene ID (Entrez) | 1490 |
| Molecular Weight (g/mol) | 36 to 44 kDa, reducing conditions |
| Quantity | 25μg |
| Source | Human embryonic kidney cell,HEK293-derived cynomolgus monkey Fc gamma RIII (CD16) protein Gly17-Gln208,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of protein by LAL method |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.