Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus IL-10 R alpha Fc Chimera Protein

Catalog No. 10186RI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-186-RI050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-186-RI050 Supplier R&D Systems™ Supplier No. 10186RI050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Interleukin-10 Receptor alpha (IL-10 R alpha), also known as IL-10 R1, is a 51 kDa transmembrane glycoprotein member of the class II cytokine receptor family

  • IL-10 R alpha is required for mediating the effects of IL-10, a critical molecule in the control of microbial infections, allergic and autoimmune inflammation, and cancer
  • IL-10 R alpha is the ligand specific subunit of the IL-10 receptor complex

Specifications

Accession Number XP_005579838.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 3587
Molecular Weight (g/mol) 51 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived cynomolgus monkey IL-10 R alpha protein

Cynomolgus Monkey IL-10 R alpha (His22-Asn235) Accession # XP_005579838.1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.