Learn More
R&D Systems™ Recombinant Cynomolgus Integrin alpha V beta 3 Protein
Description
Specifications
Specifications
| Accession Number | XP_005573729.1 (Integrin alpha V) & XP_005584667.1 (Integrin ss3) |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | M.W. Observed: 105-160 kDa, under reducing conditions, M.W. Theoretical: 115 kDa (Integrin alpha V) & 86 kDa (Integrin beta 3) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Integrin alpha V beta 3 protein Cynomolgus Monkey Integrin alpha V (Phe31-Val992) (N-terminus) Accession # XP_005573729.1 His-Pro 2 x GGGSGGGS-Acidic Tail 6-His tag Cynomolgus Monkey Integrin ss3 (Gly27-Asp718) (N-terminus) Accession # XP_005584667.1 His-Pro 2 x GGGSGGGS-Basic tail HA tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.