Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus/Rhesus IL-6R alpha His Protein

Catalog No. 11386SR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-386-SR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-386-SR050 Supplier R&D Systems™ Supplier No. 11386SR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its binding ability in a functional ELISA. Recombinant Cynomolgus Monkey/Rhesus Macaque IL-6R alpha His-tag (Catalog # 11386-SR) binds Recombinant Human IL-6 (Catalog # 7270-IL ) with an ED50 of 2.00-20.0 ng/mL.

Specifications

Accession Number XP_005541720.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3570
Molecular Weight (g/mol) MolecularWeight-observed: 60-68 kDa, under reducing conditions., MolecularWeight-theroretical: 38 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived IL-6 R alpha protein Leu20 - Asp358 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6 R-1, IL6RA, IL-6 RA, IL6RQ, interleukin 6 receptor
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-6 R alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.