Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque BCMA Fc Protein

Catalog No. 10029BC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-029-BC050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-029-BC050 Supplier R&D Systems™ Supplier No. 10029BC050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Cynomolgus/Rhesus Macaque BCMA Fc Protein is derived from HEK293. The Recombinant Cynomolgus/Rhesus Macaque BCMA Fc Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number XP_005591343
For Use With (Application) Bioactivity
Gene ID (Entrez) 608
Molecular Weight (g/mol) 33 kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived BCMA/TNFRSF17 protein, (N-terminus) Cynomolgus Monkey BCMA (Met1-Ile54) Accession &Num; XP_005591343 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Specific Reactivity BCMA/TNFRSF17
Conjugate Unconjugated
Species Cynomolgus Monkey, Rhesus macaque
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.