Learn More
R&D Systems™ Recombinant Cynomolgus SIRP beta 1/CD172b His Protein
Description
- Measured by its binding ability in a functional ELISA.
- When Recombinant Cynomolgus Monkey SIRP beta 1/CD172b is immobilized at 1 μg/mL, 100 μL/well, the concentration of Recombinant Human SP-D (Catalog # 1920-SP) that produces 50% of the optimal binding response is 0.15-0.9 μg/mL.
Specifications
Specifications
| Accession Number | XP005568593.1 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 10326 |
| Molecular Weight (g/mol) | 38 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey SIRP beta 1/CD172b protein Glu30-Pro369, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.