Learn More
R&D Systems™ Recombinant Cynomolgus VISTA/B7-H5 Fc Chimera Protein
Description
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | XP_005565644 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 64115 |
| Molecular Weight (g/mol) | M.W. (Observed): 62-71 kDa, reducing conditions; M.W. (theoretical): 45 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived cynomolgus monkey VISTA/B7-H5/PD-1 H protein Cynomolgus Monkey VISTA/B7-H5 (Phe33-Ala194) (N-terminus) Accession # XP_005565644 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.