Learn More
Abcam Recombinant Cytomegalovirus pp65 protein, 200UG
Supplier:  Abcam AB43041200UG
Encompass_Preferred
Recombinant Cytomegalovirus pp65 protein is a Human herpesvirus 5 strain AD169 Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: 65 kDa phosphoprotein, pp65, 65 kDa matrix phosphoprotein, Phosphoprotein UL83, Tegument protein UL83, UL83
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1379099