Learn More
Active Motif Recombinant dCas9 protein, His/FLAG Tag
SDP
Supplier:  Active Motif 81069
Encompass
dCas9 (deficient Cas9) protein is an endonuclease-inactive version of Cas9 (CRISPR associated protein 9). Cas9 is an RNA-guided DNA endonuclease associated with adaptive immunity system in Streptococcus pyogenes, among other bacteria. Cas9 can unwind foreign DNA and checking for sites complementary to the 20 basepair spacer region of the guide RNA. If the DNA substrate is complementary to the guide RNA, Cas9 cleaves the invading DNA. In this sense, the CRISPR-Cas9 mechanism has a number of parallels with the RNA interference (RNAi) mechanism in eukaryotes. dCas9 lacks of the endonuclease activity, so that it bind but do not cleave cognate DNA, which can be used to localize transcriptional activator or repressors to specific DNA sequences […] For more information, please visit: https://www.activemotif.com/catalog/details/81069.
Catalog No. NC1851420